missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant EGF
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
500µg
Description
- Epidermal growth factor (EGF) is characterized by high affinity binding to various EGF receptors (EGFRs) and the production of mitogenic responses (Carpenter & Cohen)
- EGF promotes EGFR dimerization, resulting in activation of downstream pathways including PI3K, ERK1/2, JAK/STAT, β-catenin, and calcium signaling
- EGF is secreted by the gut-associated salivary and Brunner's glands, is found in a variety of body fluids, and stimulates cell proliferation and differentiation in rodent and neonatal human intestine (Wright et al.) Central nervous system stem cells also proliferate in response to the EGF stimulus (Reynolds & Weiss)
Specifications
Specifications
| Name | Beta-urogastrone, HOMG4, Epidermal growth factor, Pro-epidermal growth factor, URG, Urogastrone |
| Quantity | 100 μg |
| Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | >95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction