missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-4
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Description
- Fibroblast growth factor 4 (FGF-4) is a member of the FGF superfamily (Beenken & Mohammadi)
- FGF-4 is expressed in pluripotent stem cells and is implicated in various stages of development and morphogenesis in a variety of organisms (Kosaka et al. 2009)
- FGF-4 has been shown to have an ability to promote neural stem cell proliferation and neuronal differentiation in the postnatal brain (Kosaka et al. 2006)
- FGF-4 has also been shown to increase the proliferation rate of mesenchymal stem cells (Farré et al.) FGF-4 supports the
- properties of human embryonic stem cells and also promotes the proliferation of these cells (Mayshar et al.) In the mouse, the Fgf4 gene also supports proliferation of the inner cell mass (ICM) and postimplantation embryos (Feldman et al.)
Specifications
Specifications
| Name | HBGF-4, Heparin-binding growth factor 4, Heparin secretory transforming protein 1, HST, HST-1, HSTF-1, Kaposi sarcoma oncogene, KFGF, Transforming protein KS3 |
| Quantity | 100 μg |
| Cell Type | Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction