Learn More
STEMCELL Technologies Human Recombinant FGF-4
Product Code. 17179571
Encompass_Preferred
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Product Code. 17179571
Brand: STEMCELL Technologies 78103.1
Fibroblast growth factor 4
  • Fibroblast growth factor 4 (FGF-4) is a member of the FGF superfamily (Beenken & Mohammadi)
  • FGF-4 is expressed in pluripotent stem cells and is implicated in various stages of development and morphogenesis in a variety of organisms (Kosaka et al. 2009)
  • FGF-4 has been shown to have an ability to promote neural stem cell proliferation and neuronal differentiation in the postnatal brain (Kosaka et al. 2006)
  • FGF-4 has also been shown to increase the proliferation rate of mesenchymal stem cells (Farré et al.) FGF-4 supports the
  • properties of human embryonic stem cells and also promotes the proliferation of these cells (Mayshar et al.) In the mouse, the Fgf4 gene also supports proliferation of the inner cell mass (ICM) and postimplantation embryos (Feldman et al.)
Specifications
Name HBGF-4, Heparin-binding growth factor 4, Heparin secretory transforming protein 1, HST, HST-1, HSTF-1, Kaposi sarcoma oncogene, KFGF, Transforming protein KS3
Quantity 25 μg
Cell Type Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Keratinocytes; Mammary Cells; Mesenchymal Stem and Progenitor Cells; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells; Prostate Cells
Purity or Quality Grade ≥95%