missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant GDNF, ACF
Description
- Glial cell line-derived neurotrophic factor (GDNF) is a neurotrophic factor and a member of the tumor growth factor (TGF)-beta superfamily
- The GDNF family of growth factors also includes neurturin, persephin, and artemin, which have seven conserved cysteine residues called cysteine-knot
- GDNF family ligands signal through binding to specific GDNF-family receptor-α (GFRα) co-receptors and activate the RET receptor tyrosine kinase
- Four different forms of GFRα co-receptors have been characterized (GFRα 1-4) out of which GDNF binds specifically to GFRα1 prior to forming a complex with RET
- GDNF is known to promote survival and morphological differentiation of midbrain dopaminergic neurons in both in vivo and in vitro studies and increase their high-affinity dopamine uptake
- GDNF has also been shown to have restorative effects on dying dopaminergic neurons in response to degenerative toxins
Specifications
Specifications
| Name | Astrocyte-derived trophic factor, ATF, ATF1, ATF2, Glial cell-derived neurotrophic factor |
| Quantity | 10 μg |
| Cell Type | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?