missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant HGF

Product Code. 15781846
Encompass_Preferred
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15781846

Brand: STEMCELL Technologies 78019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hepatocyte growth factor

  • Hepatocyte growth factor/scatter factor (HGF/SF) is a pleiotropic growth factor that promotes proliferation, motility, survival, and differentiation
  • HGF binds and promotes the dimerization and activation of the receptor tyrosine kinase c-MET, and stimulates PI3K/AKT, FAK, JNK, and ERK1/2 signaling (Organ & Tsao)
  • HGF stimulates migration of cells during embryogenesis, induces cell motility and scattering of epithelial cells, and regulates epithelial-mesenchymal transition
  • It is a chemoattractant for motor neurons, and regulates the development of sensory, sympathetic, parasympathetic, and cortical neurons as well as the proliferation of oligodendrocytes and glial development
  • Additionally, HGF is cytoprotective and regulates liver growth and regeneration (Nakamura et al.)

Specifications

Name DFNB39, F-TCF, Fibroblast-derived tumor cytotoxic factor, Hepapoietin A, HPTA, Lung fibroblast-derived mitogen, Scatter factor, SF
Quantity 50 μg
Cell Type Airway Cells; Hepatic Cells; Intestinal Cells; Mammary Cells; Neurons; Pluripotent Stem Cells; Prostate Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.