missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant LIF

Product Code. p-7217625
Encompass_Preferred
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 17109461

Brand: STEMCELL Technologies 78055.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Leukemia inhibitory factor

  • Leukemia inhibitory factor (LIF) is an interleukin 6 class cytokine that regulates a broad variety of developmental functions
  • After LIF binds to LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling
  • LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype
  • Produced by the endometrium, LIF plays an important autocrine and paracrine role in implantation by regulating proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment
  • Human LIF can be used for the maintenance of mouse embryonic stem cells, however mouse LIF cannot bind to the human receptor, thus rendering mouse LIF inactive
  • LIF is produced by CD4+ and activated regulatory T cells, and promotes Foxp3 expression, while repressing Th17 lineage-specific genes

Specifications

Name CDF, Cholinergic differentiation factor, D Factor, DIA, Differentiation-inducing factor, Differentiation inhibitory activity, Differentiation-stimulating factor, Emfilermin, Hepatocyte-stimulating factor III, HILDA, Leukemia inhibitory factor, MLPLI
Quantity 10 μg
Cell Type Neural Cells, PSC-Derived; Neurons; Pluripotent Stem Cells
Purity or Quality Grade >95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.