missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant M-CSF

Product Code. 16244551
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
This item is not returnable. View return policy

Product Code. 16244551

Brand: STEMCELL Technologies 78057.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Macrophage colony-stimulating factor

  • Macrophage colony-stimulating factor (M-CSF) is a homodimeric glycoprotein growth factor that regulates proliferation and differentiation of myeloid hematopoietic progenitor cells to mononuclear phagocytic cell lineages, including monocytes, macrophages, and osteoclasts
  • M-CSF is a crucial factor for the development of tissue-resident macrophages in most tissues
  • It is required for the maturation and activation of monocytes and macrophages, and regulates inflammatory responses in conjunction with other stimuli such as IFN-γ, LPS, and IL-4
  • M-CSF is also required for bone resorption by osteoclasts, and is involved in the development and regulation of the placenta, mammary gland, and brain
  • M-CSF is produced by monocytes, fibroblasts, osteoclasts, stromal cells, endothelial cells, and tumor cells

Specifications

Name Colony stimulating factor 1, CSF-1
Quantity 10 μg
Cell Type Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Monocytes; Myeloid Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.