missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant RANKL, ACF
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
Description
- Receptor activator of nuclear factor kappa-B ligand (RANKL) is a member of the tumor necrosis factor (TNF) superfamily (Anderson et al.) Cytokines in the TNF superfamily are involved in a variety of long-term cellular activities such as differentiation, proliferation, and cell death (MacEwan)
- RANKL is a type II homotrimeric transmembrane protein expressed in both a membrane-bound and secreted form (Ikeda et al.) RANKL binds to the receptor activator of nuclear factor kappa-B (RANK)
- Upon binding to its receptor, RANKL activates the AKT signaling pathway (Moon et al.) Osteoprotegerin (OPG) may also bind RANKL, and this binding competes with RANKL-RANK binding (Lacey et al.) RANKL is involved in osteoclastogenesis (Lacey et al.; Yasuda et al.) and T cell activation (Wong et al.) This product is animal component-free
Specifications
Specifications
| Name | CD254, hRANKL2, ODF, OPGL, OPTB2, Osteoclast differentiation factor, soluble Receptor activator of NF-κB ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TNF superfamily member 11, TNLG6B, Tumor necrosis factor superfamily member 11, TRAN |
| Quantity | 10 μg |
| Cell Type | Airway Cells; Cancer Cells and Cell Lines; Dendritic Cells; Intestinal Cells; Mammary Cells; Osteoblasts; Other; Pancreatic Cells; Prostate Cells; T Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction