missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reduced Folate Carrier/SLC19A1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8450U-100UG
This item is not returnable.
View return policy
Description
Reduced Folate Carrier/SLC19A1 Monoclonal specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Reduced Folate Carrier/SLC19A1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P41440 | |
| SLC19A1 | |
| NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1 Met1-Gln591 Accession # P41440 | |
| RUO | |
| 6573 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 890513 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CHMD, FLOT1, folate transporter 1, FOLT, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human Reduced Folate Carrier/SLC19A1 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction