missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reg4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1379-SP
This item is not returnable.
View return policy
Description
Reg4 Polyclonal specifically detects Reg4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Reg4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gastrointestinal secretory protein, GISP, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELP, RELPREG-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 83998 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q9BYZ8 | |
| REG4 | |
| E. coli-derived recombinant human Reg4 Asp23-Pro158 Accession # Q9BYZ8 | |
| 25 μg | |
| Primary | |
| Detects human Reg4 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse (rm) Reg4, rmReg3A, and recombinant rat (rr) Reg3A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction