Learn More
Invitrogen™ eBioscience™ Human Regulatory T Cell Staining Kit #3
Product Code. 15531237
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Contains Sodium azide and formaldehyde
Product Code. 15531237
Brand: Invitrogen™ 88899540
Contains Sodium azide and formaldehyde
Includes
  • Flow Cytometry Staining Buffer
  • Fixation/Permeabilization Concentrate (4X)
  • Fixation/Permeabilization Diluent
  • Permeabilization Buffer (10X)
  • Normal Rat Serum
  • eZFluor™ Anti-Human CD4 FITC and CD25 PE Cocktail (clones RPA-T4 and BC96, cat. 22-8425): 25 tests Light-sensitive material.
  • Anti-Human Foxp3 PE-Cyanine5 (clone PCH101, cat. 15-4776): 25 tests Light-sensitive material.
  • Rat IgG2a K Isotype Control PE-Cyanine5 : 25 μg Light-sensitive material.
Contains all of the buffers and monoclonal antibodies for CD4, CD25, and Foxp3 necessary to successfully stain and identify regulatory T cells from human peripheral blood cells.
Pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. The PCH101 antibody can be used at 20μL (0.5μg) per test.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
Quantity 1 kit
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Cell Staining Kit