Learn More
Invitrogen™ eBioscience™ Human Regulatory T Cell Whole Blood Staining Kit
Product Code. 15541237
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Contains Sodium azide and formaldehyde
Product Code. 15541237
Brand: Invitrogen™ 88899640
Contains Sodium azide and formaldehyde
Includes
  • Flow Cytometry Staining Buffer
  • 1X RBC Lysis Buffer
  • Fixation/Permeabilization Concentrate (4X)
  • Fixation/Permeabilization Diluent
  • Anti-Human Foxp3 PE (clone PCH101, cat. 12-4776): 25 tests Light-sensitive material.
Contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3+ cells in whole blood samples.
Pre-titrated and tested by flow cytometric analysis of human whole blood. The PCH101 antibody can be used at 5μL (0.25μg) per test.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
Quantity 1 kit
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Cell Staining Kit