missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RELT/TNFRSF19L Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1385N-100UG
This item is not returnable.
View return policy
Description
RELT/TNFRSF19L Monoclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for Flow Cytometry.
Specifications
| RELT/TNFRSF19L | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q969Z4 | |
| RELT | |
| Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L Met1-Ala160 (Arg127Gly, Arg129Gly) Accession # Q969Z4 | |
| RUO | |
| 84957 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 238104 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19L, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human RELT/TNFRSF19L in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, recombinant mouse (rm) 4-1BB, rhBAFF R, rmBAFF R, rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rmDR3, rhDR6, rhEDAR, rmEDAR, rhFas, rmFas, rrFas, rhGITR, rmGITR, rhHVEM, rhLT beta R, rmLT beta R, rhNGF R, rmNGF R, rhOPG, rmOPG, rmOX40, rhRANK, rmRANK, rhTNF RI, rmTNF RI, rhTNF RII, rmTNF RII, rhTRAIL R1, rhTRAIL R2, rhTRAIL R3, rhTRAIL R4, rhTROY, or rmTROY is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction