missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 160.00 - € 462.00
Specifications
| Antigen | Human Respiratory Syncytial Virus A2 Fusion Protein |
|---|---|
| Dilution | Western Blot 1:2000-1:10000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026579
|
Novus Biologicals
NBP3-26093-20ul |
20 μL |
€ 160.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30035748
|
Novus Biologicals
NBP3-26093-100ul |
100 μL |
€ 462.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Respiratory Syncytial Virus A2 Fusion Protein Polyclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western BlotSpecifications
| Human Respiratory Syncytial Virus A2 Fusion Protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| HRSVgp08, RSV, RSV fusion protein | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:2000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Rabbits immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (Uniprot#: P03420.1, Met1-Thr529) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title