missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Antibody (4-18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 372.00
Specifications
| Antigen | Human Respiratory Syncytial Virus Nucleoprotein VPN41 |
|---|---|
| Clone | 2024-04-18 00:00:00 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610678
|
Novus Biologicals
NBP2-50535-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604139
|
Novus Biologicals
NBP2-50535 |
0.1 mL |
€ 372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Nucleoprotein VPN41 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Human Respiratory Syncytial Virus Nucleoprotein VPN41 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS | |
| IgG2a | |
| Protein A purified |
| 2024-04-18 00:00:00 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Gradient-purified RSN-2 virus (subgroup B) treated with 0.1% SDS at 100C for 2 mins. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title