missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Antibody (4-18), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50535IR
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus Nucleoprotein VPN41 Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Nucleoprotein VPN41 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Human Respiratory Syncytial Virus Nucleoprotein VPN41 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Gradient-purified RSN-2 virus (subgroup B) treated with 0.1% SDS at 100C for 2 mins. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| This antibody reacts with both human RSV subgroups A and B. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA | |
| 43208 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction