Learn More
Human Respiratory Syncytial Virus Phosphoprotein Antibody (2H102), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50276R
Product Code. 18191399
  • € 466.00 / 0.10mL
Description
Human Respiratory Syncytial Virus Phosphoprotein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus Phosphoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human Respiratory Syncytial Virus Phosphoprotein
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Human RSV strain A2 infected HeLa cells
0.1 mL
Virology Bacteria and Parasites
Virus
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
2H102
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
RUO
Primary
Store at 4C in the dark.