Learn More
Human ROBO4 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB24541R-100UG
This item is not returnable.
View return policy
Description
ROBO4 Monoclonal specifically detects ROBO4 in Human samples. It is validated for Flow Cytometry.
Specifications
| ROBO4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8WZ75 | |
| ROBO4 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO4 Gln28-Arg467 Accession # Q8WZ75 | |
| RUO | |
| 54538 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 265721 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ROBO4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO2, or rhROBO3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Product Title
For Research Use Only (RUO)