missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR gamma /RORC/NR1F3 Antibody, R&D Systems™
Description
ROR gamma/RORC/NR1F3 Monoclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ROR gamma/RORC/NR1F3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1090902 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene Accession No. | P51449 |
| Gene Alias | MGC129539, NR1F3, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORC, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?