missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPRD1B (aa 72-130) Control Fragment Recombinant Protein

Product Code. 30202524
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202524

Brand: Invitrogen™ RP105387

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66832 (PA5-66832. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Interacts with phosphorylated C-terminal heptapeptide repeat domain (CTD) of the largest RNA polymerase II subunit POLR2A, and participates in dephosphorylation of the CTD by RPAP2. Transcriptional regulator which enhances expression of CCND1. Promotes binding of RNA polymerase II to the CCDN1 promoter and to the termination region before the poly-A site but decreases its binding after the poly-A site. Prevents RNA polymerase II from reading through the 3' end termination site and may allow it to be recruited back to the promoter through promotion of the formation of a chromatin loop. Also enhances the transcription of a number of other cell cycle-related genes including CDK2, CDK4, CDK6 and cyclin-E but not CDKN1A, CDKN1B or cyclin-A. Promotes cell proliferation. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQG5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 58490
Name Human RPRD1B (aa 72-130) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610304G08Rik; 2810446G03Rik; C20orf77; Cell cycle-related and expression-elevated protein in tumor; cell-cycle related and expression-elevated protein in tumor; CREPT; dJ1057B20.2; NET60; regulation of nuclear pre-mRNA domain containing 1 B; regulation of nuclear pre-mRNA domain-containing protein 1 B; RGD1304782; Rprd1b
Common Name RPRD1B
Gene Symbol RPRD1B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RKGPEFTREFESVLVDAFSHVAREADEGCKKPLERLLNIWQERSVYGGEFIQQLKLSME
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.