missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RTTN Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9966-100
This item is not returnable.
View return policy
Description
rotatin Monoclonal specifically detects rotatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| rotatin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DKFZp434G145, FLJ26356, FLJ39085, MSSP, PMGYS, rotatin | |
| Synthetic peptide containing human RTTN Accession # Q86VV8 | |
| 100 μg | |
| 25914 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 2311A | |
| Western Blot 0.5 ug/mL, ELISA, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q86VV8 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human RTTN in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction