missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4476-SP
This item is not returnable.
View return policy
Description
S100A1 Polyclonal specifically detects S100A1 in Human samples. It is validated for Western Blot.
Specifications
| S100A1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6271 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P23297 | |
| S100A1 | |
| E. coli-derived recombinant human S100A1 Met1-Ser94 Accession # P23297 | |
| 25 μg | |
| Primary | |
| Detects human S100A1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) S100B, rhS100P, and rhS100A2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction