missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4475-SP
This item is not returnable.
View return policy
Description
S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Immunohistochemistry.
Specifications
| S100A7/Psoriasin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P31151 | |
| S100A7 | |
| E. coli-derived recombinant human S100A7 Met1-Gln101 Accession # P31151 | |
| 25 μg | |
| Primary | |
| Detects human S100A7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) S100A1, A2, A4, A6, A8, A9, A10, A11, A14, A16, rhS100B, rhS100P, recombinant mouse S100A1, A4, A6, A8, A9, A10, A11, A13, or A16 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 577513 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6278 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction