missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SALM3/LRFN4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5445-SP
This item is not returnable.
View return policy
Description
SALM3/LRFN4 Monoclonal specifically detects SALM3/LRFN4 in Human samples. It is validated for Immunohistochemistry.
Specifications
| SALM3/LRFN4 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q6PJG9 | |
| LRFN4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SALM3/LRFN4 Cys17-Leu518 Accession # Q6PJG9 | |
| 25 μg | |
| Primary | |
| Detects human SALM3/LRFN4 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) SALM1, approximately 20-25% cross-reactivity with recombinant human rhSALM2, and no cross-reactivity with rhSALM4 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 578102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIGLER6, LRFN4, SALM3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 78999 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction