missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SAPAP4 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
SAPAP4 Synthetic Peptide, PEP-0678, from Invitrogen.
SAP90/PSD-95-associated protein 4 (SAPAP4, also known as DLGAP4) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP4 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP4 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -2, whereas SAPAP3 mRNA is detected mainly in cell bodies.
Specifications
Specifications
| Gene ID (Entrez) | 22839 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9Y2H0 |
| Common Name | Human SAPAP4 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction