missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SCF/c-kit Ligand Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF255
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| SCF/c-kit Ligand | |
| Polyclonal | |
| Biotin | |
| P21583 | |
| KITLG | |
| E. coli-derived recombinant human SCF/c-kit Ligand (R&D Systems, Catalog # 255-SC) Glu26-Ala189 Accession # P21583 | |
| 50 μg | |
| Primary | |
| Detects human SCF/c-kit Ligand in ELISAs and Western blots. In sandwich immunoassays, approximately 55% cross-reactivity with recombinant feline SCF is observed, 20% cross-reactivity with recombinant canine SCF is observed, and less than 0.3% cross-reactivity with recombinant mouse SCF is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4254 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction