missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Semaphorin 3B (NP_004627.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | Semaphorin 3 B |
| Accession Number | NP_004627.1 |
| Gene ID (Entrez) | 7869 |
| Gene Alias | (semaphorin) 3 B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, SEMA3B, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3 B, Semaphorin-V, SEMAV |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction