missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 4B Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF5485
This item is not returnable.
View return policy
Description
Semaphorin 4B Polyclonal specifically detects Semaphorin 4B in Human samples. It is validated for Western Blot.
Specifications
| Semaphorin 4B | |
| Polyclonal | |
| Biotin | |
| Q9NPR2 | |
| SEMA4B | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4B isoform 1 Leu39-Glu712 Accession # Q9NPR2 | |
| 50 μg | |
| Primary | |
| Detects human Semaphorin 4B in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Semaphorin 4B is observed and less than 1% cross-reactivity with recombinant human Semaphorin 3A, 4A, and 7A is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| KIAA1745sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4B, MGC131831, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4B, SEMA4B, SEMAC, semaphorin-4B, SemC | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10509 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction