missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 7A Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB20681V-100UG
This item is not returnable.
View return policy
Description
Semaphorin 7A Monoclonal specifically detects Semaphorin 7A in Human samples. It is validated for Flow Cytometry.
Specifications
| Semaphorin 7A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O75326 | |
| SEMA7A | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 7A His47-Ala648 Accession # O75326 | |
| RUO | |
| 8482 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 310829 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD108, CD108 antigen, CD108MGC126696, CDw108, H-SEMA-K1, H-Sema-L, JMH, JMH blood group antigen, John-Milton-Hargen human blood group Ag, MGC126692, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A, sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin)7A (JMH blood group), sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A, Sema K1, Sema L, Sema7A, SEMAK1, SEMAL, SEMALJohn Milton Hagen blood group H-Sema K1, semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group), semaphorin K1, semaphorin L, semaphorin-7A, semaphorin-K1, Semaphorin-L | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Semaphorin 7A in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction