missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A5/Protein C Inhibitor Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1266G-100UG
This item is not returnable.
View return policy
Description
Serpin A5/Protein C Inhibitor Monoclonal specifically detects Serpin A5/Protein C Inhibitor in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Serpin A5/Protein C Inhibitor | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Acrosomal serine protease inhibitor, antitrypsin), member 5, member 5, PAI-3, PAI3PLANH3, PCI, PCIplasminogen activator inhibitor III, Plasminogen activator inhibitor 3, plasminogen activator inhibitor-3, PROCIplasma serine protease inhibitor, Protein C inhibitor, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A5, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), SERPNA5 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A5/Protein C, His20-Pro406, Accession # P05154 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 210326 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/106 cells | |
| P05154 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5104 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction