missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A7/TBG Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8176-SP
This item is not returnable.
View return policy
Description
Serpin A7/TBG Monoclonal specifically detects Serpin A7/TBG in Human samples. It is validated for Western Blot.
Specifications
| Serpin A7/TBG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P05543 | |
| SERPINA7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Serpin A7/TBG Met1-Ala415 Accession # P05543 | |
| 25 μg | |
| Primary | |
| Detects human Serpin A7/TBG in ELISA and Western Blot. | |
| Human | |
| Purified |
| Western Blot | |
| 893339 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBG, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6906 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction