missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A9/Centerin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5429-SP
This item is not returnable.
View return policy
Description
Serpin A9/Centerin Polyclonal specifically detects Serpin A9/Centerin in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation.
Specifications
| Serpin A9/Centerin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Centerin, GCET1, SERPINA11, SERPINA11b | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 327657 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAQ89063 | |
| SERPINA9 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A9 Ala24-Thr417 Accession # AAQ89063 | |
| 25 μg | |
| Primary | |
| Detects human Serpin A9 in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction