missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serpin B3/SCCA1 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human Serpin B3/SCCA1 quantitates human Serpin B3/SCCA1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin B3/SCCA1.
May act as a papain-like cysteine protease inhibitor to modulate the host immune response against tumor cells. Also functions as an inhibitor of UV-induced apoptosis via suppression of the activity of c-Jun NH(2)-terminal kinase (JNK1). [UniProt]
Specifications
Specifications
| Accession Number | HsT1196; Protein T4-A; SCC; SCCA; SCCA1; SCCA-1; SCCA-PD; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; serine protease inhibitor B3; serpin B3; serpin B4; serpin family B member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; serpin peptidase inhibitor, clade B (ovalbumin), member 4; Serpinb3; SERPINB4; spb3; Squamous cell carcinoma antigen 1; SSCA1; T4-A |
| Assay Range | 0.156 to 10 ng/mL |
| Assay Sensitivity | 0.15 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6317 |
| Gene Symbol | SERPINB3 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction