missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin F2/alpha 2-Antiplasmin Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF1470
This item is not returnable.
View return policy
Description
Serpin F2/alpha 2-Antiplasmin Polyclonal specifically detects Serpin F2/alpha 2-Antiplasmin in Human samples. It is validated for Western Blot.
Specifications
| Serpin F2/alpha 2-Antiplasmin | |
| Polyclonal | |
| Biotin | |
| P08697 | |
| SERPINF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin F2/alpha 2 -Antiplasmin Met28-Lys491 Accession # P08697 | |
| 50 μg | |
| Primary | |
| Detects human Serpin F2/alpha 2 -Antiplasmin in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin A4, rhSerpin A5, recombinant mouse (rm) Serpin C1 and rmSerpin D1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5345 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction