missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1455T-100UG
This item is not returnable.
View return policy
Description
Albumin Monoclonal specifically detects Albumin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Albumin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Serum Albumin in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 188835 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ALB | |
| Human Serum Albumin | |
| RUO | |
| 213 | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction