missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Antibody (10C7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 187.00
Specifications
| Antigen | Human Serum Albumin |
|---|---|
| Clone | 10C7 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
Description
Human Serum Albumin Monoclonal specifically detects Human Serum Albumin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmunoassay.Specifications
| Human Serum Albumin | |
| 1 mg/mL | |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| RUO | |
| Human | |
| albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| ALB | |
| IgG2a | |
| Affinity Purified |
| 10C7 | |
| ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| P02768 | |
| 213 | |
| Human Serum Albumin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title