missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21669
This item is not returnable.
View return policy
Description
Human Serum Albumin Polyclonal specifically detects Human Serum Albumin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Human Serum Albumin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from Antiserum with 0.01% Sodium Azide | |
| albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Rabbit | |
| 69.37 kDa | |
| 2 mL | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 213 | |
| Reconstitute with 2.0 ml deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| P02768 | |
| ALB | |
| Albumin (Human Serum) (Uniprot: P02768) | |
| Delipidation and Defibrination | |
| RUO | |
| Primary | |
| Cross reactivity against Albumin from other tissues and species may occur but has not been specifically determined. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction