missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 Alexa Fluor™ 350-conjugated Antibody, Novus Biologicals™

Human Monoclonal Antibody
Brand: R&D Systems LFAB10733U-100UG
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal antibody specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
| Mouse myeloma cell line CHO-derived human Siglec-3/CD33, Val18-His259, Accession # AAA51948 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| L008.2.14N | |
| Flow Cytometry | |
| AAA51948 | |
| Human | |
| His-tag purified from cell culture supernatant | |
| RUO | |
| 945 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction