missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 208.00 - € 2100.00
Specifications
| Antigen | Siglec-3/CD33 |
|---|---|
| Clone | 6C5/2 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751647
![]() |
R&D Systems
MAB1137-SP |
25 μg |
€ 208.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792937
![]() |
R&D Systems
MAB1137-100 |
100 μg |
€ 520.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761127
![]() |
R&D Systems
MAB1137-500 |
500 μg |
€ 2100.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Siglec-3/CD33 | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 945 | |
| Recombinant human Siglec-3/CD33 Extracellular domain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Siglec-3/CD33 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -5, -7, or -9 is observed. |
| 6C5/2 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
| CD33 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title