missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-3/CD33 (Research Grade Gemtuzumab Biosimilar) Alexa Fluor 750-conjugated Antibody, R&D Systems™
Human Monoclonal Antibody
Brand: R&D Systems FAB9635S-100UG
This item is not returnable.
View return policy
Description
Siglec-3/CD33 Monoclonal specifically detects Siglec-3/CD33 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-3/CD33 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Siglec-3/CD33 based on Gemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Gemtuzumab. This product is for research use only. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu9a | |
| Flow Cytometry 0.25-1μg/106 cells | |
| CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67 | |
| Human Siglec-3/CD33 | |
| 100 μg | |
| 945 | |
| Human | |
| Supernatant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur