missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-5/Siglec-14 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10721R-100UG
This item is not returnable.
View return policy
Description
Siglec-5/Siglec-14 Monoclonal specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Flow Cytometry.
Specifications
| Siglec-5/Siglec-14 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15389 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Siglec-5/Siglec-14 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant human (rh) Siglec-14 is observed and no cross-reactivity with rhSiglec-3, rhSiglec-7, or rhSiglec-9 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 194128 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| SIGLEC5 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-5/Siglec-14 Lys18-Thr434 Accession # O15389 | |
| RUO | |
| 8778 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction