missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-5/Siglec-14 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF1072
This item is not returnable.
View return policy
Description
Siglec-5/Siglec-14 Polyclonal specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Siglec-5/Siglec-14 | |
| Polyclonal | |
| Biotin | |
| O15389 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8778 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| SIGLEC5 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-5 Lys18-Thr434 Accession # O15389 | |
| 50 μg | |
| Primary | |
| Detects human Siglec-5/Siglec-14 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Siglec-7 and rhSiglec-9 is observed and less than 5% cross-reactivity with rhSiglec-2 and rhSiglec-3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction