missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SIK2 (aa 325-457) Control Fragment Recombinant Protein

Product Code. 30206759
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206759

Brand: Invitrogen™ RP91454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82546 (PA5-82546. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase that belongs to the AMP-activated protein kinase family. SNF1LK2 negatively regulates CRE-binding protein (CREB) activity by phosphorylating the CREB-specific coactivator transducer of regulated CREB activity (TORC). SIK2 is thought to be part of a signaling cascade that regulates the expression and activity of the insulin-induced genes PGC-1'alpha and UCP-1 in brown adipocytes, impairment of which has been implicated in obesity and insulin resistance in human and animal models. SIK2 has also been reported as a key regulator for neuronal survival after ischemia, suppressing CREB-mediated gene expression after oxygen-glucose deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H0K1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23235
Name Human SIK2 (aa 325-457) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias G630080D20Rik; KIAA0781; LOH11CR1I; QIK; Qin-induced kinase; RGD1559698; salt induceable kinase 2; salt inducible kinase 2; salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; Sik2; SIK-2; SNF1-like kinase 2; SNF1LK2
Common Name SIK2
Gene Symbol SIK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HFAAIYFLLVERLKSHRSSFPVEQRLDGRQRRPSTIAEQTVAKAQTVGLPVTMHSPNMRLLRSALLPQASNVEAFSFPASGCQAEAAFMEEECVDTPKVNGCLLDPVPPVLVRKGCQSLPSNMMETSIDEGLE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.