Learn More
Gibco™ Human IL-12 Receptor beta-1 (soluble) Fc Recombinant Protein, PeproTech®
Description
Recombinant Human sIL-12 Receptor Beta-1 Fc has a calculated molecular weight of 84.3 kDa and contains 758 amino acid residues, which consists of the extracellular domain of IL-12R Beta-1 fused to the Fc portion of human IgG and contains a C-terminal His-tag. Due to glycosylation, it migrates at an approximate molecular weight of 116 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P42701 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 3594 |
| Molecular Weight (g/mol) | 84.3 kDa |
| Name | Human IL-12 Receptor beta-1 (soluble) Fc |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.