missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SIRP beta 1/gamma Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF2096
This item is not returnable.
View return policy
Description
SIRP beta 1/gamma Polyclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Western Blot.
Specifications
| SIRP beta 1/gamma | |
| Polyclonal | |
| Biotin | |
| O00241 | |
| SIRPB1 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241 | |
| 50 μg | |
| Primary | |
| Detects human SIRP beta 1 and human SIRP gamma in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10326 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction