missing translation for 'onlineSavingsMsg'
Learn More

Human SIRP gamma/CD172g Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18750068
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18750068 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18750068 Supplier R&D Systems Supplier No. MAB4486SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP gamma/CD172g Monoclonal specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP gamma/CD172g
Applications Western Blot
Classification Monoclonal
Clone 595337
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9P1W8
Gene Alias bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-b2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPG, SIRPgamma, SIRP-gamma
Gene Symbols SIRPG
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SIRP gamma/CD172g Val64-Ser364 Accession # Q9P1W8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55423
Test Specificity Detects human SIRP gamma/CD172g in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human (rh) SIRP alpha and rhSIRP beta 1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.