missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sirtuin 1/SIRT1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7714
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Sirtuin 1/SIRT1 | |
| Monoclonal | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q96EB6 | |
| SIRT1 | |
| E. coli-derived recombinant human Sirtuin 1/SIRT1 Met1-Ser747 Accession # Q96EB6 | |
| 100 μg | |
| Primary | |
| Detects human Sirtuin 1/SIRT1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Sirtuin 2, 3, 5, or 6 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 834918 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.5.1, S. cerevisiae, homolog) 1, SIR2alpha, SIR2L1, SIR2-like protein 1, SIRT1, sirtuin 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23411 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction