missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sirtuin 1/SIRT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7714-SP
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated.
Specifications
| Sirtuin 1/SIRT1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.5.1, S. cerevisiae, homolog) 1, SIR2alpha, SIR2L1, SIR2-like protein 1, SIRT1, sirtuin 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23411 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF, KnockDown | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated | |
| Q96EB6 | |
| SIRT1 | |
| E. coli-derived recombinant human Sirtuin 1/SIRT1 Ala2-Ser747 Accession # Q96EB6 | |
| 25 μg | |
| Primary | |
| Detects human Sirtuin 1/SIRT1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SIRT2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction