missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLFN14 Control Fragment Recombinant Protein

Product Code. 30208168
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208168

Brand: Invitrogen™ RP93543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54684 (PA5-54684. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Despite being first described several years ago, the roles of the Schlafen (SLFN) family of proteins remain largely unknown. The SLFN genes are preferentially expressed in lymphoid tissues and differentially regulated during thymocyte maturation. It is thought that many play roles in cell growth, hemopoietic cell differentiation, and T cell development and maturation. Most members contain at least one divergent AAA domain (AAA_4) that may play a role in ATP binding. SLFN14 is a recently identified member of the SLFN family; its role has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P0C7P3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 342618
Name Human SLFN14 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BDPLT20; C-terminally truncated SLFN14 endoribonuclease; Gm20; LRRGT00163; Protein SLFN14; schlafen 14; schlafen family member 14; schlafen family member 14, pseudogene; SLFN14; Slfn14-ps
Common Name SLFN14
Gene Symbol SLFN14
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DPFQIHHADVNGLPPPSAQFPRKTITSGIHCALEIAKVMKEEMKRIKENPPSNMSPDTLALFSETAYEEATCAQALPGVCETKTNLTTEQIANYVGRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.