missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3009-SP
This item is not returnable.
View return policy
Description
SLITRK1 Monoclonal specifically detects SLITRK1 in Human samples. It is validated for Western Blot.
Specifications
| SLITRK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96PX8 | |
| SLITRK1 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK1 Ala16-Ser616 Accession # Q96PX8 | |
| 25 μg | |
| Primary | |
| Detects human SLITRK1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, or rhSLITRK5 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 327808 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 114798 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction